Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
Thermo Scientific Chemicals

Silica gel, for column chrom., ultra pure, 35-70 µm, 60A

For column chromatography. | CAS: 7631-86-9
数量:
1 kg
5 kg
30 kg
製品番号(カタログ番号) 360050010
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
さらに表示
pH6.5 to 7.0 (5 % suspension in water)
Particle size=<10 % (Undersize)
Appearance (Form)Powder
Specific surface area460 to 520 m²/g
Particle size35 to 70 µm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only