Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Thermo Scientific Chemicals

Silica gel, for column chrom., ultrapure, 60-200 micron, 60A

CAS: 7631-86-9 | O2Si | 60.08 g/mol
数量:
250 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic Drum
製品番号(カタログ番号) 360060050
価格(JPY)
-
見積もりを依頼する
数量:
5 kg
パッケージ:
Plastic Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
さらに表示
Appearance (Color)White
Appearance (Form)Powder
Specific surface area470 to 530 m²/g
Pore diameter58 to 68 A
Pore volume0.70 to 0.85 ml/g
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
RUO – Research Use Only