Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron
Thermo Scientific Chemicals

Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 360110500
Price (USD)
447.65
Online Exclusive
497.00
Save 49.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
447.65
Online Exclusive
497.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS71889-02-6
IUPAC Namesilanedione; trichloro(octadecyl)silane
Molecular FormulaC18H37Cl3O2Si2
InChI KeyVYTRPUAMVCWPLO-UHFFFAOYSA-N
SMILESO=[Si]=O.CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Specific surface area480 to 550 m²/g
Carbon (C)>=20.7 %
Volatile matter=<4 % (110°C)
Pore volume0.70 to 0.85 ml/g
Loading>=0.96 mmol/g (Molecular)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 μm (Cat. No. 360110500, 360112500) in Angstroms?

The pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 µm (Cat. No. 360110500, 360112500) is 60 Angstroms.