Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron
Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron
Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron
Thermo Scientific Chemicals

Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron

CAS: 71888-97-6 | C9H23NO5Si2 | 281.455 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Catalog number 360180250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS71888-97-6
IUPAC Name(3-aminopropyl)triethoxysilane; silanedione
Molecular FormulaC9H23NO5Si2
InChI KeyJIBSXWZIQTVOKJ-UHFFFAOYSA-N
SMILESO=[Si]=O.CCO[Si](CCCN)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loading>=1.2 mmol/g (molecular)
Nitrogen (N)>=1.68 %
Appearance (Color)White to off-white
Particle size40 to 63 µm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only