Sodium carbonate monohydrate, 99.5-100.5% (by anhydrous basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium carbonate monohydrate, 99.5-100.5% (by anhydrous basis), crystals, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF, JP, BP and EP specifications.
Have Questions?
變更視圖buttonViewtableView
Quantity:
2.5 kg
12 kg
100 lb.
產品號碼 3603-05
價格 (HKD)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets NF, JP, BP and EP specifications.
This product is GMP Manufactured and meets NF, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
規格
ColorWhite
Melting Point107.85°C
PackagingPoly bottle, tamper-evident cap
GradeMulti-Compendial, NF
Molecular Weight (g/mol)124
Percent Purity99.5 to 100.5%
Physical FormSolid
Quantity2.5 kg
Unit Size2.5 kg