4-Bromomethylbenzenesulfonyl chloride, 95%
4-Bromomethylbenzenesulfonyl chloride, 95%
4-Bromomethylbenzenesulfonyl chloride, 95%
Thermo Scientific Chemicals

4-Bromomethylbenzenesulfonyl chloride, 95%

CAS: 66176-39-4 | C7H6BrClO2S | 269.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 360540250
Price (USD)
332.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
332.00
Each
Chemical Identifiers
CAS66176-39-4
IUPAC Name4-(bromomethyl)benzene-1-sulfonyl chloride
Molecular FormulaC7H6BrClO2S
InChI KeyQXTQWYZHHMQSQH-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point72°C to 77°C
Titration Argentometric>=94 %
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only