Sodium carbonate, 99.5-100.5% (by anhydrous basis), NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium carbonate, 99.5-100.5% (by anhydrous basis), NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets NF, JP, BP, and EP specifications.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
100 lb.
200 lb.
Número de catálogo 3606-01
Precio (MXN)
-
Cantidad:
500 g
This product is GMP Manufactured and meets NF, JP, BP, and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorBlanco
Punto de fusión854°C
EnvaseAmber glass bottle, wide mouth, tamper-evident cap
Punto de ebullición1600°C (1013 hPa)
GradoMulti-Compendial, NF
Peso molecular (g/mol)105.99
Porcentaje de pureza99.5 to 100.5%
Forma físicaSolid
Cantidad500 g
pH11.4 to 11.7 (25°C)
Unit Size500 g