Specifications
ColorWhite
Melting Point854°C
PackagingPolyethylene pail, screw top
Boiling Point1600°C (1013 hPa)
GradeMulti-Compendial, NF
Molecular Weight (g/mol)105.99
Percent Purity99.5 to 100.5%
Physical FormSolid
Quantity2.5 kg
pH11.4 to 11.7 (25°C)
Unit Size2.5 kg