Specifications
ColorWhite
Melting Point854°C
PackagingPolyethylene drum, open head
Boiling Point1600°C (1013 hPa)
GradeMulti-Compendial, NF
Molecular Weight (g/mol)105.99
Percent Purity99.5 to 100.5%
Physical FormSolid
Quantity100 lb.
pH11.4 to 11.7 (25°C)
Unit SizeEach