5-Bromo-6-chloro-2-picoline, 98+%, Thermo Scientific Chemicals
5-Bromo-6-chloro-2-picoline, 98+%, Thermo Scientific Chemicals
5-Bromo-6-chloro-2-picoline, 98+%, Thermo Scientific Chemicals
5-Bromo-6-chloro-2-picoline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-6-chloro-2-picoline, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
3608000505 gGlass bottle
Catalog number 360800050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS185017-72-5
IUPAC Name3-bromo-2-chloro-6-methylpyridine
Molecular FormulaC6H5BrClN
InChI KeyJVDQYSIJBRTRMS-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
GC>=98.0 %
Appearance (Form)Low melting solid or liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only