Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack

Safely store and ship light-sensitive, small-volume, diagnostic samples and valuable reagents with these opaque, amber HDPE bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
362004-905015 mL20-415 mm
362004-91254 mL13-415 mm
362004-90258 mL20-415 mm
Catalog number 362004-9050
Price (USD)
860.00
Online Exclusive
1,075.00
Save 215.00 (20%)
Case of 2000
Capacity (Metric):
15 mL
Neck Finish:
20-415 mm
Price (USD)
860.00
Online Exclusive
1,075.00
Save 215.00 (20%)
Case of 2000

Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Durable, uniform walls resistant to splitting or puncturing
  • Leakproof† when used with Nalgene closures (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. Packed 2,000 per case. Order colored closures separately (Cat. No. 362150).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.5 oz., 15 mL
Diameter (Metric)25 mm
Diameter (Metric) Inner Neck14 mm
Height (English)2.28 in.
Height (Metric)58 mm
Neck Finish20-415 mm
No. per Case2000
No. per Pack2000
Through the Closure DispensingNo
Width (English)0.98 in.
Capacity (English)0.5 oz.
Capacity (Metric)15 mL
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure IncludedYes
ColorOpaque Amber
MaterialHDPE
MouthNarrow
Product TypeSample Container
SterilityNon-sterile
Unit SizeCase of 2000