Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Thermo Scientific™

Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with these HDPE diagnostic bottles without closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck Finish
362008-90258 mL20-415 mm
362008-91254 mL13-415 mm
362008-905015 mL20-415 mm
Catalog number 362008-9025
Price (KRW)
1,059,000
Case of 2000
Capacity (Metric):
8 mL
Neck Finish:
20-415 mm
Price (KRW)
1,059,000
Case of 2000

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure. Safe and leakproof† when used with Nalgene closures, these bottles can also be used in test kits and many other small-volume applications. Bottles are made of opaque white, high-density polyethylene and are packed without closures. They can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof† when assembled with Nalgene closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. 2000 per case. Order colored closures separately (Cat. No. 362150).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Capacity0.25 oz., 8 mL
Capacity (English)0.25 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure MaterialPolypropylene
Closure Size20-415 mm
Diameter (Metric)25 mm
Diameter (Metric) Inner Neck14 mm
Height (English)1.77 in.
Height (Metric)45 mm
Neck Finish20-415 mm
No. per Case2000
No. per Pack2000
Product TypeSample Container
ShapeRound
Width (English)0.98 in.
Capacity (Metric)8 mL
Closure IncludedNo
ColorOpaque White
GraduatedNo
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 2000