Sodium chloride, 99.0-100.5% (dried basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Have Questions?
보기 방식 변경buttonViewtableView
수량:
500 g
2.5 kg
12 kg
100 lb.
50 kg
350 lb.
카탈로그 번호 3627-05
제품 가격(KRW)
-
수량:
2.5 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
사양
끓는점1413°C (1013 hPa)
CAS7647-14-5
화학물질 이름 또는 재질Sodium chloride
색상White
용융점801°C
분자식NaCl
동의어Halite, Rock salt, Salt, Sea salt
등급Multi-Compendial
분자량(g/mol)58.44
백분율 순도99.0-100.5%
수량2.5 kg
Unit SizeEach