Sodium chloride, 99.0-100.5% (dried basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
100 lb.
50 kg
350 lb.
Catalog number 3627-08
Price (MXN)
-
Quantity:
350 lb.
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Specifications
Boiling Point1413°C (1013 hPa)
CAS7647-14-5
Chemical Name or MaterialSodium chloride
ColorWhite
Melting Point801°C
Molecular FormulaNaCl
SynonymHalite, Saline, Salt, Sea salt
Molecular Weight (g/mol)58.44
Percent Purity99.0-100.5%
Quantity350 lb.
Unit SizeEach