Sodium chloride, 99.0-100.5% (dry basis), granular, FCC, USP, GMP, J.T.Baker™
Avantor™

Sodium chloride, 99.0-100.5% (dry basis), granular, FCC, USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets F.C.C. and U.S.P Requirements.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
100 lb.
50 kg
350 lb.
Packaging:
Ver tudo
Polyethylene drum, open head
Número do catálogo 3628-20
Preço (BRL)
-
Quantity:
100 lb.
Packaging:
Polyethylene drum, open head
This product is GMP Manufactured and meets F.C.C. and U.S.P Requirements.
This product is GMP Manufactured and meets F.C.C. and U.S.P Requirements.
Especificações
Boiling Point1413°C (1013 hPa)
CAS7647-14-5
Chemical Name or MaterialSodium chloride
ColorWhite
Melting Point801°C
Molecular FormulaNaCl
PackagingPolyethylene drum, open head
SynonymHalite, Rock salt, Saline
GradeFCC
Molecular Weight (g/mol)58.44
Percent Purity99.0-100.5%
Quantity100 lb.
Unit SizeEach