Sodium chloride, 99.0-100.5% (dry basis), granular, FCC, USP, GMP, J.T.Baker™
Avantor™

Sodium chloride, 99.0-100.5% (dry basis), granular, FCC, USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets F.C.C. and U.S.P Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
100 lb.
50 kg
350 lb.
Envase:
Ver todo
Pail, HDPE Flowmor™ package
Polyethylene pail, screw top
Número de catálogo 3628-F7
Precio (MXN)
-
Cantidad:
12 kg
Envase:
Pail, HDPE Flowmor™ package
This product is GMP Manufactured and meets F.C.C. and U.S.P Requirements.
This product is GMP Manufactured and meets F.C.C. and U.S.P Requirements.
Especificaciones
Punto de ebullición1413°C (1013 hPa)
Nombre del producto químico o materialSodium chloride
ColorBlanco
Punto de fusión801°C
Fórmula molecularNaCl
EnvasePail, HDPE Flowmor™ package
SinónimoHalite, Rock salt, Saline, Salt, Sea salt
Peso molecular (g/mol)58.44
Porcentaje de pureza99.0-100.5%
Cantidad12 kg
Unit SizeEach