Nalgene™ Amber PPCO High-Profile Closures with Color Coders for Micro Packaging Vials, orange - FAQs
Nalgene™ Amber PPCO High-Profile Closures with Color Coders for Micro Packaging Vials, orange - FAQs
View additional product information for Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile - FAQs (362825-0115, 362825-0116, 362825-0113, 362825-0114, 362825-0111, 362825-0112, 362825-1110, 362825-1111, 362825-0119, 362825-0117, 362825-0118)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.