Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Thermo Scientific™

Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile

Identify samples quickly with these amber PPCO high-profile closures with color-coded inserts. Eliminate risky O-rings while providing a barrier for light-sensitive reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
362825-1111Amber
362825-0111White
362825-0112Yellow
362825-0113Orange
362825-0114Green
362825-0115Red
362825-0116Blue
362825-0117Gold
362825-0118Purple
362825-0119Natural
362825-1110Teal
Catalog number 362825-1111
Price (USD)
312.78
Online Exclusive
367.98
Save 55.20 (15%)
Case of 1000
Color:
Amber
Price (USD)
312.78
Online Exclusive
367.98
Save 55.20 (15%)
Case of 1000

Identify samples quickly using Thermo Scientific™ Nalgene™ Amber PPCO High-Profile Closures with Color Coders for Micro Packaging Vials. Noncytotoxic and nonpyrogenic, these closures do not use O-rings that might fall out or contaminate contents.

Features:

  • Vials (listed separately) and closures are pressure-tested together at 7.5psig.
  • Screw closure is threaded.
  • Permanently attached, color-coded inserts are free of toxic heavy metals.
  • Inserts do not come in contact with vial contents.
  • Create a leakproof† seal when used with Nalgene Micro Packaging Vials

Ordering Information:
Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1000 per case. Order nonsterile amber vials separately (Cat. No. 362805).

WARNING: Cancer – www.P65Warnings.ca.gov

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Certifications/ComplianceISO 13485:2016
Closure MaterialPPCO
Closure Size11 mm
DescriptionHigh-profile closures w/color coders for micro packaging vials
No. per Case1000
No. per Pack500
Product TypeClosure
Closure TypeScrew Cap
ColorAmber
Diameter (Metric) Outer11 mm
MaterialPPCO
Product LineNalgene
Quantity500 Bag, 1000 Cs.
SterilityNon-sterile
Unit SizeCase of 1000

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.