(R)-2-Isobutilsuccínico ácido-1-metil éster, 95 %, (98 % EE), Thermo Scientific Chemicals
(R)-2-Isobutilsuccínico ácido-1-metil éster, 95 %, (98 % EE), Thermo Scientific Chemicals
(R)-2-Isobutilsuccínico ácido-1-metil éster, 95 %, (98 % EE), Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-2-Isobutilsuccínico ácido-1-metil éster, 95 %, (98 % EE), Thermo Scientific Chemicals

CAS: 130165-76-3 | C9H16O4 | 188.22 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mg
1 g
Envase:
Frasco de vidrio
Número de catálogo 363050010
Precio (USD)
-
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS130165-76-3
IUPAC Name(3R)-3-(methoxycarbonyl)-5-methylhexanoate
Molecular FormulaC9H15O4
InChI KeyAJNNFMRCMKGYHA-SSDOTTSWSA-M
SMILESCOC(=O)[C@H](CC(C)C)CC([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=94 %
Enantiomeric excess>=97.5 % (GC)
Appearance (Color)Yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only