(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals

数量:
500 mg
5 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 363065000
価格(JPY)
-
数量:
500 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS220498-08-8
IUPAC Name(3R)-3-(methoxycarbonyl)-4-methylpentanoate
Molecular FormulaC8H13O4
InChI KeyVHOGSWBAPDPWTG-ZCFIWIBFSA-M
SMILESCOC(=O)[C@H](CC([O-])=O)C(C)C
さらに表示
Appearance (Form)Viscous liquid
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=94 %
Enantiomeric excess>=97.5 % (GC)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only