Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%
Thermo Scientific Chemicals

Di-μ-chlorobis(p-cymene)chlororuthenium(II), 98%

CAS: 52462-29-0 | C20H28Cl4Ru2 | 612.39 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 363290050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS52462-29-0
IUPAC Namebis(1-methyl-4-(propan-2-yl)benzene); bis(dichlororuthenium)
Molecular FormulaC20H28Cl4Ru2
InChI KeyLAXRNWSASWOFOT-UHFFFAOYSA-J
SMILESCl[Ru]Cl.Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.CC(C)C1=CC=C(C)C=C1
さらに表示
Appearance (Color)Orange-red to red-brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Elemental Analysis>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only