Thermo Scientific Chemicals

Hydrogen hexachloroplatinate(IV) hydrate, 99.9%, (trace metal basis), 38 to 40% Pt, Thermo Scientific Chemicals

Catalog number: 363590050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydrogen hexachloroplatinate(IV) hydrate, 99.9%, (trace metal basis), 38 to 40% Pt, Thermo Scientific Chemicals

Catalog number: 363590050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 363590050
Price (USD)
Price: 575.00
Your price:
Quantity
-

Chemical Identifiers

CAS
26023-84-7
IUPAC Name
dihydrogen hexachloroplatinumtetrakis(ylium)
Molecular Formula
Cl6H2Pt
InChI Key
ZKOQTCCVSRSECD-UHFFFAOYSA-J
SMILES
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
Appearance (Color)
Orange to orange-brown
Total trace metal impurities
=<0.1 %
Solubility
(1 g in 10 ml ethanol) Clear orange
Appearance (Form)
Powder and/or chunks
Assay
38 to 40 % (Pt)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text