Tris(hydroxymethyl)phosphine, 95%
Tris(hydroxymethyl)phosphine, 95%
Tris(hydroxymethyl)phosphine, 95%
Thermo Scientific Chemicals

Tris(hydroxymethyl)phosphine, 95%

CAS: 2767-80-8 | C3H9O3P | 124.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Packaging:
Glass bottle
Catalog number 363840250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2767-80-8
IUPAC Name[bis(hydroxymethyl)phosphanyl]methanol
Molecular FormulaC3H9O3P
InChI KeyJMXMXKRNIYCNRV-UHFFFAOYSA-N
SMILESOCP(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
NMR>=94 %
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only