Dicarbonylacetylacetonato rhodium(I)
Dicarbonylacetylacetonato rhodium(I)
Dicarbonylacetylacetonato rhodium(I)
Thermo Scientific Chemicals

Dicarbonylacetylacetonato rhodium(I)

CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number 363950025
Price (EUR)
2.426,00
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
2.426,00
Each
Chemical Identifiers
CAS14874-82-9
IUPAC Nameλ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC7H7O4Rh
InChI KeyBZCAWKOPWNIDOC-FGSKAQBVSA-M
SMILES[Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green-red
Appearance (Form)Flakes or needles
Rhodium (Rh)>=39.3 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only