Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 364311000
Price (EUR)
72,65
Online exclusive
81,10
Save 8,45 (10%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
72,65
Online exclusive
81,10
Save 8,45 (10%)
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.001 %
GC>=99.85 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only