Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 364351000
Price (JPY)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.9 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only