n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 364361000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only