Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 67-56-1 | CH4O | 32.04 g/mol
Quantity:
100 mL
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 364390025
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
GC>=99.8 %
Acidity/alkalinity=<0.0005 meq/g
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only