Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 1L, Vapor Pressure: 20mbar at 20 deg.C, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL | CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 364420010
Price (USD)
210.65
Online exclusive
234.00
Save 23.35
Each
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
210.65
Online exclusive
234.00
Save 23.35
Each
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Water=<0.005 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only