Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 364661000
Price (USD)
26.65
Online exclusive
29.30
Save 2.65
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
26.65
Online exclusive
29.30
Save 2.65
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=99.4 %
Acidity=<0.001 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only