Trisodium citrate dihydrate, ≥99% (by anhydrous basis), granular, BP, Ph. Eur., JP, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Trisodium citrate dihydrate, ≥99% (by anhydrous basis), granular, BP, Ph. Eur., JP, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P, and E.P Specifications.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
100 lb.
Número de catálogo 3647-20
Precio (EUR)
-
Cantidad:
100 lb.
This product is GMP Manufactured and meets J.P., U.S.P, B.P, and E.P Specifications.
Especificaciones
CAS4-3-6132
Nombre del producto químico o materialTrisodium citrate
ColorBlanco
Punto de fusión150°C
Fórmula molecularHOC(COONa)(CH2COONa)2·2H2O
EnvasePolyethylene drum, open head
SinónimoCitric acid tridosium salt dihydrate, Sodium citrate dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate
GradoMulti-Compendial
Peso molecular (g/mol)294.1
Porcentaje de pureza≥99%
Cantidad100 lb.
Unit SizeEach