Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals
Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals
Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals

CAS: 4418-26-2 | Molecular Formula: C8H7O4Na | Molecular Weight: 190.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
364855000500 gPlastic bottle
Catalog number 364855000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS4418-26-2
IUPAC Namesodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide
Molecular FormulaC8H7NaO4
InChI KeyYIOCQGHBBNGBND-UHFFFAOYSA-N
SMILES[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Titration with HClO4>=98.5 %
Appearance (Form)Powder

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only