Benzyltrimethylammonium bromide, 98+%
Benzyltrimethylammonium bromide, 98+%
Benzyltrimethylammonium bromide, 98+%
Thermo Scientific Chemicals

Benzyltrimethylammonium bromide, 98+%

CAS: 5350-41-4 | C10H16BrN | 230.15 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 364861000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5350-41-4
IUPAC Namebenzyltrimethylazanium bromide
Molecular FormulaC10H16BrN
InChI KeyUUZYBYIOAZTMGC-UHFFFAOYSA-M
SMILES[Br-].C[N+](C)(C)CC1=CC=CC=C1
さらに表示
Appearance (Form)Crystalline powder
Appearance (Color)White to cream
Infrared spectrumConforms
Melting point224°C to 234°C
Titration Argentometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only