Trisodium citrate dihydrate, 99.0-100.5% (by anhydrous basis), granular, FCC, USP, GMP, J.T.Baker™
Avantor™

Trisodium citrate dihydrate, 99.0-100.5% (by anhydrous basis), granular, FCC, USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
50 kg
100 kg
Catalog number 3649-08
Price (MXN)
-
Quantity:
100 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Specifications
CAS4-3-6132
Chemical Name or MaterialTrisodium Citrate
ColorWhite
Melting Point150°C
Molecular FormulaNa3C6H5O7·2H2O
PackagingPolyethylene drum, open head
SynonymSodium Citrate Dihydrate, Citric acid trisodium salt dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Citric acid tridosium salt dihydrate
GradeFCC
Molecular Weight (g/mol)294.1
Percent Purity99.0-100.5%
Quantity100 kg
Unit SizeEach