6-Hydroxyquinoline, 98%
6-Hydroxyquinoline, 98%
6-Hydroxyquinoline, 98%
Thermo Scientific Chemicals

6-Hydroxyquinoline, 98%

CAS: 580-16-5 | C9H7NO | 145.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 364930250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS580-16-5
IUPAC Namequinolin-6-ol
Molecular FormulaC9H7NO
InChI KeyOVYWMEWYEJLIER-UHFFFAOYSA-N
SMILESOC1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Beige to grey to brown
HPLC>=97.5 %
Melting point190°C to 195°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only