Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Thermo Scientific™

Plunger-in-Needle Autosampler Syringes for Thermo Scientific™ Instruments

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.
Have Questions?
Catalog NumberNeedle Gauge
3650404626 G
Catalog number 36504046
Price (USD)
193.35
Pack of 1
Needle Gauge:
26 G
Price (USD)
193.35
Pack of 1

Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Thermo Scientific Instruments.

  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
Specifications
Description0.5uL volume; 26 gauge; 80mm length
DisposableReusable
Length (Metric) Needle80 mm
Needle Point StyleCone
Product TypeGC Autosampler Syringe, Plunger-in-Needle
For Use With (Equipment)TriPlus Autosampler
Needle Gauge26 G
Volume (Metric)0.5 μL
Unit SizePack of 1