L(+)-Arabinose, 99%, for biochemistry
L(+)-Arabinose, 99%, for biochemistry
L(+)-Arabinose, 99%, for biochemistry
Thermo Scientific Chemicals

L(+)-Arabinose, 99%, for biochemistry

CAS: 87-72-9 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 365181000
Price (USD)
225.65
Online exclusive
251.00
Save 25.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
225.65
Online exclusive
251.00
Save 25.35
Each
Chemical Identifiers
CAS87-72-9
IUPAC Name(3S,4R,5R)-oxane-2,3,4,5-tetrol
Molecular FormulaC5H10O5
InChI KeySRBFZHDQGSBBOR-ZRMNMSDTSA-N
SMILESO[C@@H]1COC(O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Specific optical rotation+102° to +106° (20°C, 589 nm) (c=10, water)
Residue after ign. (sulfated)=<0.3 %
Heavy metals (as Pb)=<10 ppm
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only