Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™

Removable-Needle Autosampler Syringes for GC Instruments

Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeQuantityNeedle Point Style
36520051250 μL23 GEa.Side Hole
365C09215 μL26 GEa.Cone
365C09265 μL26 G6/Pk.Cone
36500495100 μL23 GEa.Cone
36520050100 μL23 GEa.Side Hole
3650052010 μL31 GEa.Cone
365G150350 μL23 GEa.Cone
Catalog number 36520051
Price (MXN)
-
Volume (Metric):
250 μL
Needle Gauge:
23 G
Quantity:
Ea.
Needle Point Style:
Side Hole
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionUse when performing PTV/LVI injections with a dedicated liner for thermal labile compounds.; 36520051
DisposableReusable
Length (Metric)50 mm
Length (Metric) Needle50 mm
Needle Point StyleSide Hole
Product TypeGas Tight GC Autosampler Syringe, Removable Needle
QuantityEa.
Replacement Needle36550040
Connection TypeRemovable
For Use With (Equipment)TriPlus, AS2000, AS200/AS800 Autosampler
Needle Gauge23 G
Volume (Metric)250 μL
Unit SizeEach