Thermo Scientific Chemicals

2,3-Dichlorobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: 365580050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

2,3-Dichlorobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: 365580050
5 g, Glass Bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
57915-78-3
IUPAC Name
1-(bromomethyl)-2,3-dichlorobenzene
Molecular Formula
C7H5BrCl2
InChI Key
PVIGUZZDWGININ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(CBr)=C1Cl
Appearance (Color)
White
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
GC
>=98.5 %
Melting point
29°C to 32°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text