Ethanol, 95+%, pure, denat. with 5% wood spirit
Ethanol, 95+%, pure, denat. with 5% wood spirit
Ethanol, 95+%, pure, denat. with 5% wood spirit
Ethanol, 95+%, pure, denat. with 5% wood spirit
Thermo Scientific Chemicals

Ethanol, 95+%, pure, denat. with 5% wood spirit

CAS: 64-17-5 | C2H6O | 46.07 g/mol
数量:
1 L
2.5 L
10 L
製品番号(カタログ番号) 365780100
価格(JPY)
-
見積もりを依頼する
数量:
10 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
さらに表示
Appearance (Color)Clear colorless
GC=<5 % (wood spirit)
GC>=95.0 % (ethanol)
Appearance (Form)Liquid
Refractive index1.3600 to 1.3630 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use