Removable-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Thermo Scientific™

Removable-Needle Autosampler Syringes for Thermo Scientific™ Instruments

Use Thermo Scientific™ Removable-Needle Syringes for accurate and reproducible sample introduction into GC instruments, with or without the needle.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeLength (Metric)For Use With (Equipment)
365D161110 μL23 G42 mmAgilent GC Instruments
365D184110 μL26 G50 mmCTC GC Instruments
365D373110 μL23 G50 mmCTC GC Instruments
3650301550 μL23 G50 mmAS2000 Autosampler
365B02511 μL23 G57 mmTriPlus RSH Autosampler
365D661010 μL26s G42 mmShimadzu GC Instruments
Catalog number 365D1611
Price (USD)
-
Volume (Metric):
10 μL
Needle Gauge:
23 G
Length (Metric):
42 mm
For Use With (Equipment):
Agilent GC Instruments

Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Removable-Needle Syringes offer accurate and reproducible sample introduction, with or without the needle. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.

  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DisposableReusable
Length (Metric)42 mm
Length (Metric) Needle42 mm
Needle Point StyleCone
Product TypeGC Autosampler Syringe, Removable Needle
For Use With (Equipment)Agilent GC Instruments
Needle Gauge23 G
Volume (Metric)10 μL
Unit SizeEach