Thermo Scientific™
Repair Kits for Thermo Scientific™ Plunger-in-Needle Syringes
 Related applications:
Industrial Chromatography
Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Repair kits for plunger in needle syringes contain both the plunger and needle, which must be changed together
Includes: Plunger, 1; Needle, 1
Documents
Manuals & protocols