Nalgene™ Plastic Graduated Cylinder Variety Pack
Nalgene™ Plastic Graduated Cylinder Variety Pack
Nalgene™ Plastic Graduated Cylinder Variety Pack
Nalgene™ Plastic Graduated Cylinder Variety Pack
Thermo Scientific™

Nalgene™ Plastic Graduated Cylinder Variety Pack

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Seven sizes, from 10 to 1000mL.

Have Questions?
Número do catálogoCapacity (Metric)
3662-123410, 25, 50, 100, 250, 500, 1000 mL
Número do catálogo 3662-1234
Preço (BRL)
-
Capacity (Metric):
10, 25, 50, 100, 250, 500, 1000 mL

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Thermo Scientific™ Nalgene™ Plastic Graduated Cylinder Variety Packs include one each 10mL, 25mL, 50mL, 100mL, 250mL, 500mL and 1000mL cylinders. All sizes have a generous pour spout and molded-in graduations that are inked for easy reading. No meniscus allows accurate readings every time. The large base keeps cylinders stable on the benchtop preventing tipping hazards—won't break or chip like glass labware. Suitable for food and beverage use.

  • Includes seven cylinders, one each size: 10mL, 25mL, 50mL, 100mL, 250mL, 500mL and 1000mL
  • Guaranteed to meet requirements of ASTM Class B, E1272 for accuracy
  • No meniscus to confuse readings—eliminates guesswork
  • Large blue octagonal base prevents rolling and tipping
  • Generous pour spout provides ease in dispensing liquids
  • Molded-in graduations are silk-screened for easy reading
  • Excellent chemical resistance for a wide variety of solutions
  • Impact resistant at temperatures above -40°C
  • Autoclavable/Graduated
Compliance:
  • Meets ASTM Class B, E1272 Class B and 21 CFR Part 177.1520 for food/beverage use
Notes:
  • Autoclaving will affect graduation accuracy
Especificações
AutoclavableAutoclavable
Certifications/ComplianceASTM, CFR 21 Part 177.1520, E1272
For Use With (Application)Food and Beverage Use, Refreshing an Existing Lab
IncludesBlue Polypropylene Base
Max. Temperature (English)250°F
Max. Temperature (Metric)121°C
Quantity7/Pk.
ScalemL
ShapeRound
SubdivisionYes
Base TypeOctagonal
Capacity (Metric)10, 25, 50, 100, 250, 500, 1000 mL
MaterialPPCO
Product TypeGraduated Cylinder Set
Unit SizeEach

Frequently asked questions (FAQs)

Do you offer class A plastic graduated cylinders?

We do not offer plastic graduated cylinders with Class A accuracy. It is very difficult to reach Class A accuracy with plastic, which is why they are almost always glass.