D(+)-Fenchone, 97%
D(+)-Fenchone, 97%
D(+)-Fenchone, 97%
Thermo Scientific Chemicals

D(+)-Fenchone, 97%

CAS: 4695-62-9 | C10H16O | 152.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
366291000100 gGlass bottle
Catalog number 366291000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range96% min. (GC)
Beilstein07, IV, 212
CAS4695-62-9
ChEBICHEBI:165
Chemical Name or MaterialD(+)-Fenchone
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only