4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%

CAS: 269409-70-3 | C12H17BO3 | 220.08 g/mol
Have Questions?
Catalog NumberQuantity
3664200505 g
Catalog number 366420050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS269409-70-3
IUPAC Name4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular FormulaC12H17BO3
InChI KeyBICZJRAGTCRORZ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light brown
Appearance (Form)Crystalline powder
GC>=96.0 %
Melting point112°C to 117°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only