1,2,3,4-Butanetetracarboxylic acid, 99+%
1,2,3,4-Butanetetracarboxylic acid, 99+%
1,2,3,4-Butanetetracarboxylic acid, 99+%
Thermo Scientific Chemicals

1,2,3,4-Butanetetracarboxylic acid, 99+%

CAS: 1703-58-8 | C8H10O8 | 234.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 366761000
Price (USD)
52.65
Online exclusive
58.00
Save 5.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.65
Online exclusive
58.00
Save 5.35
Each
Chemical Identifiers
CAS1703-58-8
IUPAC Namebutane-1,2,3,4-tetracarboxylic acid
Molecular FormulaC8H10O8
InChI KeyGGAUUQHSCNMCAU-UHFFFAOYNA-N
SMILESOC(=O)CC(C(CC(O)=O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point188°C to 192°C
Titration with NaOH99.0 to 101.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only