Anhídrido p-toluenosulfónico, + 95 %, Thermo Scientific Chemicals
Anhídrido p-toluenosulfónico, + 95 %, Thermo Scientific Chemicals
Anhídrido p-toluenosulfónico, + 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anhídrido p-toluenosulfónico, + 95 %, Thermo Scientific Chemicals

CAS: 4124-41-8 | C14H14O5S2 | 326.39 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 367070250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS4124-41-8
IUPAC Name4-methylbenzenesulfonyl 4-methylbenzene-1-sulfonate
Molecular FormulaC14H14O5S2
InChI KeyPDVFSPNIEOYOQL-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C1=CC=C(C)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Titration by coupling>=94 %
Appearance (Color)Off-white to beige to grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only