4-N-BOC-Aminopiperidine, 98%
4-N-BOC-Aminopiperidine, 98%
4-N-BOC-Aminopiperidine, 98%
Thermo Scientific Chemicals

4-N-BOC-Aminopiperidine, 98%

CAS: 73874-95-0 | C10H21N2O2 | 201.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 367620250
Price (EUR)
431,00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
431,00
Each
Chemical Identifiers
CAS73874-95-0
IUPAC Name4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium
Molecular FormulaC10H21N2O2
InChI KeyCKXZPVPIDOJLLM-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)NC1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Melting point155°C to 160°C
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only