2,3-Dichlorophenylboronic acid, 97%
2,3-Dichlorophenylboronic acid, 97%
2,3-Dichlorophenylboronic acid, 97%
Thermo Scientific Chemicals

2,3-Dichlorophenylboronic acid, 97%

CAS: 151169-74-3 | C6H5BCl2O2 | 190.81 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3677100505 gGlass bottle
Catalog number 367710050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS151169-74-3
IUPAC Name(2,3-dichlorophenyl)boronic acid
Molecular FormulaC6H5BCl2O2
InChI KeyTYIKXPOMOYDGCS-UHFFFAOYSA-N
SMILESOB(O)C1=C(Cl)C(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
HPLC>=96.0 %
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only