Quinoline-3-boronic acid, 97%
Quinoline-3-boronic acid, 97%
Quinoline-3-boronic acid, 97%
Thermo Scientific Chemicals

Quinoline-3-boronic acid, 97%

CAS: 191162-39-7 | C9H8BNO2 | 172.98 g/mol
数量:
1 g
5 g
25 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 367730050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS191162-39-7
IUPAC Name(quinolin-3-yl)boronic acid
Molecular FormulaC9H8BNO2
InChI KeyYGDICLRMNDWZAK-UHFFFAOYSA-N
SMILESOB(O)C1=CC2=CC=CC=C2N=C1
さらに表示
Titration with NaOH>=96.0 %
Appearance (Form)Powder
Appearance (Color)Light pink to orange to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only